Learn More
TPGS2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA559225
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: TFTAYYRLLI THLGLPQWQY AFTSYGISPQ AKQWFSMYKP ITYNTNLLTE ETDSFVNKLD PSKVFKSKNK IVIPK Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
TPGS2 Polyclonal antibody specifically detects TPGS2 in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
TPGS2 | |
Polyclonal | |
Unconjugated | |
TPGS2 | |
C18orf10; HMFN0601; HsT3006; L17; PGs2; TPGS2; Tubulin polyglutamylase complex subunit 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25941 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q68CL5 | |
TPGS2 | |
Recombinant protein corresponding to Human TPGS2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only