Learn More
TFF2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA557781
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: PHNRTNCGFP GITSDQCFDN GCCFDSSVTG VPWCFHPLPK QESDQCVMEV SDRRNCGYPG ISPEECASRK CCFSNFIFEV PWC Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 83%.
Trefoil peptides are protease resistant molecules secreted throughout the gut that play a role in mucosal healing and protection of the gastrointestinal epithelia. These peptides contain three intrachain disulfide bonds, forming the trefoil motif, or P-domain. SP (spasmolytic polypeptide), also designated Trefoil factor 2 (TFF2) precursor, is a trefoil protein that functions to inhibit gastrointestinal motility and gastric acid secretion. SP may also act as a structural component of the gastric mucus, possibly by stabilizing glycoproteins in the mucus gel through interactions with carbohydrate side chains. A downregulation of SP expression is associated with primary gastric cancer, and a progressive loss of this protein is likely to be involved in the early stage of the multi-step gastric carcinogenesis pathway.Specifications
TFF2 | |
Polyclonal | |
Unconjugated | |
Tff2 | |
mSP; pancreatic spasmolytic polypeptide; PSP; SML1; SP; spasmolysin; Spasmolytic polypeptide; spasmolytic protein 1; Tff2; Trefoil factor 2; trefoil factor 2 (spasmolytic protein 1) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7032 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.10 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q03403 | |
Tff2 | |
Recombinant protein corresponding to Human TFF2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only