missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydropyran-2-ylmethylamine, 97%, Thermo Scientific Chemicals

Catalog Number p-7077418
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
6628-83-7
C6H13NO
115.17
NYGCBQKDTGBHSC-UHFFFAOYSA-N
tetrahydropyran-2-ylmethylamine, 2-aminomethyl tetrahydropyran, tetrahydro-2h-pyran-2-yl methanamine, 2-aminomethyl tetrahydro-2h-pyran, 2h-pyran-2-methanamine, tetrahydro, 2-aminomethyltetrahydropyran, c-tetrahydro-pyran-2-yl-methylamine, pyran, 2-aminomethyl tetrahydro, pyran, 2-aminomethyltetrahydro, oxan-2-yl methanamine
97924
oxan-2-ylmethanamine
C1CCOC(C1)CN
This item is not returnable. View return policy
6628-83-7
C6H13NO
115.17
NYGCBQKDTGBHSC-UHFFFAOYSA-N
tetrahydropyran-2-ylmethylamine, 2-aminomethyl tetrahydropyran, tetrahydro-2h-pyran-2-yl methanamine, 2-aminomethyl tetrahydro-2h-pyran, 2h-pyran-2-methanamine, tetrahydro, 2-aminomethyltetrahydropyran, c-tetrahydro-pyran-2-yl-methylamine, pyran, 2-aminomethyl tetrahydro, pyran, 2-aminomethyltetrahydro, oxan-2-yl methanamine
97924
oxan-2-ylmethanamine
C1CCOC(C1)CN

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 110.0°C (25.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 115.17
Percent Purity 97%
Chemical Name or Material Tetrahydropyran-2-ylmethylamine
compliance-icons
Product Identifier
  • Tetrahydropyran-2-ylmethylamine, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'