missing translation for 'onlineSavingsMsg'
Learn More

Terazosin hydrochloride dihydrate, 98%, Thermo Scientific Chemicals

Catalog Number p-7077858
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
70024-40-7
ClH·2H2O
459.92
NZMOFYDMGFQZLS-UHFFFAOYSA-N
terazosin hydrochloride dihydrate, magnurol, dysalfa, flotrin, heitrin, hytrine, adecur, deflox, urodie, hytrin
63016
CHEBI:9446
[4-(4-amino-6,7-dimethoxyquinazolin-2-yl)piperazin-1-yl]-(oxolan-2-yl)methanone;dihydrate;hydrochloride
COC1=C(C=C2C(=C1)C(=NC(=N2)N3CCN(CC3)C(=O)C4CCCO4)N)OC.O.O.Cl
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
70024-40-7
ClH·2H2O
459.92
NZMOFYDMGFQZLS-UHFFFAOYSA-N
terazosin hydrochloride dihydrate, magnurol, dysalfa, flotrin, heitrin, hytrine, adecur, deflox, urodie, hytrin
63016
CHEBI:9446
[4-(4-amino-6,7-dimethoxyquinazolin-2-yl)piperazin-1-yl]-(oxolan-2-yl)methanone;dihydrate;hydrochloride
COC1=C(C=C2C(=C1)C(=NC(=N2)N3CCN(CC3)C(=O)C4CCCO4)N)OC.O.O.Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 271.0°C to 274.0°C
Loss on Drying 9% max. (105°C, 3 h)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 459.92
Percent Purity 98%
Chemical Name or Material Terazosin hydrochloride dihydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'