Learn More
TCR alpha/beta Monoclonal Antibody (IP26), eFluor™ 450, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
150.00 € - 346.00 €
Specifications
Antigen | TCR alpha/beta |
---|---|
Clone | IP26 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16646384
|
Affymetrix eBioscience
48-9986-42 |
100 Tests |
346.00 €
100 tests |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16636384
|
Affymetrix eBioscience
48-9986-41 |
25 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The IP26 monoclonal antibody reacts with the alpha beta chain of human TCR. The alpha beta TCR is expressed by the majority of peripheral T cells. Applications Reported: This IP26 antibody has been reported for use in flow cytometric analysis. Applications Tested: This IP26 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor 450 is an alternative to Pacific Blue. eFluor 450 emits at 446 nm and is excited with the violet laser line (405 nm). Please make sure that your instrument is capable of detecting this fluorochrome. Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser
TCR alpha/beta Monoclonal antibody specifically detects TCR alpha/beta in Human samples. It is validated for Flow Cytometry.Specifications
TCR alpha/beta | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
28589, 28755, 6955, 6957 | |
IgG1 κ | |
Affinity chromatography |
IP26 | |
Flow Cytometry | |
eFluor 450 | |
Mouse | |
Human | |
P01848, P04435 | |
TRA, TRAC, TRB, TRBV7-9 | |
Primary | |
Antibody |