Learn More
Synuclein Ab-2 Mouse Monoclonal Antibody, Epredia™
Provide accurate, reproducible results Epredia™ Synuclein Ab-2, Mouse Monoclonal Antibody.
112.00 € - 635.00 €
Specifications
Antigen | Synuclein Ab-2 |
---|---|
Clone | syn211 |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
12669996
|
Epredia
MS-1572-PABX |
Concentrated | 200 μL |
635.00 €
200µL |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12649996
|
Epredia
MS-1572-P1 |
Concentrated | 500 μL |
389.00 €
500µL |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12639996
|
Epredia
MS-1572-P0 |
Concentrated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12659996
|
Epredia
MS-1572-P1ABX |
Concentrated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12629996
|
Epredia
MS-1572-P |
Concentrated | 1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12689996
|
Epredia
MS-1572-R7 |
Prediluted | 7 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Synnuclein family of proteins is abundantly expressed in neuronal cytosol and presynaptic terminals. In vertebrates they are encoded by three different genes. Synucleins have been specifically implemented in three major diseases: Alzheimer's (AD), Parkinson's (PD) and breast cancer. In AD, a peptide derived from α-synuclein forms an intrinsic component of plaque amyloid. In PD, an alpha synuclein accumulates in Lewy bodies. An allele of α synuclein has been linked to many familial cases of PD. In breast cancer increased expression of γ synuclein correlates with the disease progression. Synucleins appear to be involved in the membrane plasticity in developing song control system of songbirds.Host Species: Mouse
Clone: syn211
Isotype: IgG1
Species Reactivity: Human. Does not react with mouse and rat. Others not known.
Epitope: Not determined
Immunogen: Human recombinant α-synuclein
Molecular Weight: ∽14kDa
Positive Control: Brain
Cellular Localization: Neuronal cytosol and presynaptic terminals
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation
Note:
USA: RUO; Int'l: RUO
Specifications
Synuclein Ab-2 | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
RUO | |
Human | |
IgG1 |
syn211 | |
Monoclonal | |
Mouse | |
Neuroscience | |
Human recombinant alpha-synuclein | |
Primary |