Learn More
Miele Professional™ Cover
Filter E 319/3 for optimum protection against contamination from labels or glass splinters
Brand: Miele Professional™ 6477070
Additional Details : Weight : 0.50000kg
Description
- Surface filter for coarse waste
- Prevents soiling by labelling laboratory glassware
- Prevents contamination with splinters of broken glass
Specifications
Stainless Steel | |
Stainless Steel | |
Washer-disinfectors, laboratory | |
500 mm |
Cover | |
E 319/3 Insert 1/1 | |
488 mm | |
1 Each |