missing translation for 'onlineSavingsMsg'
Learn More

Labcon SuperSlik™ 10 μL Low Retention Pipette Tips

Ultra low fluid retention

Brand:  Labcon 1162-965-006-9

Additional Details : Weight : 13.15420kg

 View more versions of this product

Product Code. 16651943

  • 74.05 € / Pack of 960

Estimated Shipment: 18-02-2022
Add to basket

Description

Description

  • Pipette tip
  • Graduated at 2 μL and 10 μL
  • With Ultrafine™ point
  • Fits Popular 10 μL Pipettors
  • Ultra low fluid retention
  • Protease, Human DNA, ATP, RNase/DNase and Endotoxin Free (non-pyrogenic)
  • Polypropylene USP Class VI resin
  • Made in USA
Specifications

Specifications

Yes
Polypropylene USP Class VI resin
10 μL
Sterile
Purple
1.50 in.
ISO 9001:2015; FDA CFR21 GMP; CE IVD Conformity
Liquid Handling
Beveled
96 tips / rack
No
SuperSlik™ Low Retention
Non-Filtered
Natural
Autoclavable
38.1 mm
Protease, Human DNA, ATP, RNase/DNase and Endotoxin Free (non-pyrogenic)
Fits Popular 10 µL Pipettors
96 tips/rack; 10 racks / Pk.; 10 Pk. / case
RT
Videos
SDS
Documents
Certificates
Promotions

Promotions