Learn More
Labcon SuperSlik™ 10 μL Low Retention Pipette Tips
Ultra low fluid retention
Brand:  Labcon 1161-265-000-9
Additional Details : Weight : 5.44310kg
 View more versions of this product
Product Code. 16653982
  • 50.39 € / Pack of 1248
Estimated Shipment: 11-02-2022
Add to basket
Description
Description
  • Pipette Tip
  • Graduated at 2 μL and 10 μL
  • With UltraFine™ point
  • Fits Popular 10 μL Pipettors
  • Ultra low fluid retention
  • Protease, Human DNA, ATP, RNase/DNase and Endotoxin Free (non-pyrogenic)
  • Polypropylene USP Class VI resin
  • Made in USA
Specifications
Specifications
Yes
2, 10 µL
SuperSlik™ Low Retention
Non-Filtered
Clear
1.23 in.
ISO 9001:2015; FDA CFR21 GMP; CE IVD Conformity
Liquid Handing
Fits Popular 10 µL Pipettors
1248 tips / low waste Eclipse™ refill/10 refills / case
Yes
Polypropylene USP Class VI resin
10 μL
Sterile
Autoclavable
31.4 mm
Protease, Human DNA, ATP, RNase/DNase and Endotoxin Free (non-pyrogenic)
E-Beam
Graduated, UltraFine tip
1248 tips / low waste Eclipse™ refill