Learn More
Invitrogen™ eBioscience™ Streptavidin APC-eFluor™ 780 Conjugate
Commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry. Streptavidin binds to biotin with high affinity.
Brand:  Invitrogen™ 47-4317-82
Additional Details : Weight : 0.25000kg
Product Code. 15341650
  • 214.00 € / 100µg
In Stock
to see stock.
Add to basket
Alert:
Contains Azide
Specifications
0.2 mg/mL
eBioscience™
Biotin
Liquid
Streptavidin Conjugate (Fluorescent)
APC-eFluor 780