missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ STK3 (MST2) Recombinant Human Protein

STK3 (MST2) is a STE kinase family member that acts upstream of the MAP kinase signaling pathways.

1826.00 € - 7890.00 €

Specifications

Accession Number NP_006272.2
Formulation 50mM Tris pH 7.5, 150mM NaCl, 0.02% Trition X-100, 0.5mM EDTA, 2mM DTT, 50% Glycerol
Molecular Weight (g/mol) 85.5kDa
Storage Requirements Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Regulatory Status RUO - research use only
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15418693
View Documents Promotion Details
Invitrogen™
PR8881A
100μg
1826.00 €
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 17-04-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15418763
View Documents Promotion Details
Invitrogen™
PV4806
1mg
7890.00 €
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 17-04-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

STK3 (MST2) is known to be activated by the proapoptotic stimuli staurosporine and FAS ligand.

Specifications

Specifications

NP_006272.2
85.5kDa
RUO - research use only
STK3 (MST2)
Human
GST Tag
Frozen
50mM Tris pH 7.5, 150mM NaCl, 0.02% Trition X-100, 0.5mM EDTA, 2mM DTT, 50% Glycerol
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
STE20 Kinase Family
Recombinant
Baculovirus
Videos
SDS
Documents

Documents

Product Certifications