missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STK25 (YSK1) Recombinant Human Protein
STK25 (YSK1) Recombinant Human Protein
Brand: Invitrogen PV3657
Additional Details : Weight : 0.05250kg
Description
STK25 (YSK1) is a serine⁄threonine kinase that belongs to the GCK family of kinases. It has been implicated as an upstream regulator of MAP kinase signaling pathways.Specifications
NP_006365 | |
Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
75.6kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
STK25 (YSK1) | |
STK25 (YSK1) was pre-diluted in enzyme dilution buffer and assayed in 12.5mM Tris (pH 7.5), 10mM MgCl2, 1mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 667μg/mL myelin basic protein (MBP) and trace [32P]-γ-ATP for 10 minutes at 30°C | |
STK25 (YSK1) Recombinant Human Protein | |
Human | |
Baculovirus | |
Serine/threonine kinase |
0.37mg/mL | |
10494 | |
STK25 (YSK1) Recombinant Human Protein | |
–80°C | |
1 Each | |
STK25 (YSK1) Recombinant Human Protein | |
STE20 | |
320nmoles of phosphate transferred to myelin basic protein (MBP) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 3.33μg/mL | |
GST Tag | |
Kinase and inhibitors | |
95% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.