Learn More
Fritsch™ Stainless Steel Test Sieves
Brand:  Fritsch™ 30.3800.03
Additional Details : Weight : 0.30000kg
 View more versions of this product
Product Code. 10014900
  • 155.00 € / Each
Estimated Shipment: 03-04-2023
to see stock.
Add to basket
Specifications
Test Sieve
Stainless Steel
50 mm
500 μm
ISO 3310-1
200 mm
1 Ea.