Learn More
Ohaus™ Stainless Steel Calibration Weights - OIML F2
Adjusted to Meet or Exceed OIML F2 Tolerances
Brand:  Ohaus™ 30390855
Additional Details : Weight : 10.00000kg
 View more versions of this product
Product Code. 16652222
  • 535.00 € / Each
Estimated Shipment: 03-06-2024
to see stock.
Add to basket
Description
Description
  • Weights in masses ranging from 1 gram to 1 kilogram are made of Troemner Alloy 8 Stainless Steel which possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion
  • Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free, protecting your investment
  • Weight surfaces meet or exceed OIML R 111 and have a highly polished mirror-like finish which allows scratches and foreign matters on the weight to be easily seen allowing for stability and longevity
Specifications
Specifications
No Certificate
10 kg
Protective Case
Stainless Steel (316)
160 mg