missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ SSPN Recombinant Protein
POA
Specifications
Cross Reactivity | Human |
---|---|
Formulation | 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer |
Molecular Weight (g/mol) | 49.5 |
Name | SSPN (Human) Recombinant Protein (P01) |
pH Range | 8 |
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
Human | |
49.5 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
Yes | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
SSPN (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) |