Learn More
3M™ Specialty Disposable Unvalved Respirator
Provides lightweight, effective, comfortable and hygienic respiratory FFP1 protection against dust, mist and nuisance-level acid gases. 3M™ Specialty Disposable Unvalved FFP1 Respirators have a robust shell and cup design which conforms well to most face shapes and sizes and is collapse resistant.
Brand:  3M™ 7000034737
11.55 EUR valid until 2024-03-30
Use promo code "21636" to get your promotional price.
Additional Details : Weight : 0.34380kg
Product Code. 12362399
  • 120.05 € / Pack of 20
Estimated Shipment: 22-04-2024
to see stock.
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Alert:
Due to extraordinary demand patterns, this item may experience longer lead times and shipping/delivery dates may not be accurate.
Description
Description
  • Activated carbon layer
  • Convex shape, nose clip and twin-strap design
  • Maximum usage level: up to 4 x TLV for particulates; below TLV for acid gases