Learn More
Macherey-Nagel™ SPE Column Kit
Brand: Macherey-Nagel™ 730125
Additional Details : Weight : 0.01000kg
Specifications
SPE | |
SPE Analysis of PCBs | |
25 columns each of Chromabond SiOH-H2 SO4/SA and Chromabond SiOH |
Solid Phase Extraction Analysis of PCBs | |
Solid Phase Extration Column Kit |