Learn More
Poulten & Graf GmbH Spare Piston for FORTUNA™ OPTIFIX™ SAFETY Dispenser
Poulten & Graf GmbH Spare Piston for FORTUNA™ OPTIFIX™ SAFETY Dispenser comes with a finger guard and volume setting system.
Brand:  Poulten & Graf GmbH 101 00338
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10086091
  • 269.00 € / Each
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
Description
Description
  • Solid PTFE encased
  • With finger guard and volume setting system
  • With sealing lip
Specifications
Specifications
10 mL
10 mL
1
Spare part for bottle top dispenser
Reusable
Piston for OPTIFIX SAFETY
OPTIFIX SAFETY Bottle top dispenser
1
Not Autoclavable