missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Wide Mouth Color-Code Safety Labeled Wash Bottles

Catalog Number p-3273594
Click to view available options
Capacity:
16 oz., 500 mL
For Use With (Application):
Ethyl Acetate
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

Labeling with Department of Transportation (DOT) and National Fire Protection Association (NFPA) codes and symbols, Chemical Abstract Service (CAS) number, protective equipment symbols and target organ information

Easy-to-read labeling printed bottle promoting safe working environment

  • Wide mouth 53mm (2 in.) bottle for easy refilling
  • Made from solvent-resistant low-density polyethylene
  • Polypropylene closure with leakproof design
  • Pictogram instructions promote universal safety regardless of setting

Use LYOB only with solvents compatible with LDPE.

Compliance

Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200

Capacity 16 oz., 500 mL
Closure Material Polypropylene
Closure Size 53 mm
Capacity (Metric) 500 mL
Capacity (English) 16 oz.
Color Clear
For Use With (Application) Ethyl Acetate
Material LDPE
Autoclavable Not Autoclavable
Mouth Wide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'