missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Nitrite, ≥99.0%, Reagent Grade, Honeywell™
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly bottle
Description
- ≥95% for general laboratory use.
Specifications
Melting Point | 280°C |
Color | Colorless to Yellow |
pH | 8 to 9 (at 100.00g/L 20°C) |
Quantity | 500 g |
Linear Formula | NaNO2 |
UN Number | UN1500 |
Formula Weight | 69g/mol |
Percent Purity | ≥99.0% |
Ignition Point | 914°F |
Odor | Odorless |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'