missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydride, ca 60% dispersion in oil, in solvent solublebags, Thermo Scientific™

Catalog Number p-7052550
Click to view available options
Quantity:
50 g
100 g
500 g
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
7646-69-7
HNa
23.998
MFCD00003471
BZKBCQXYZZXSCO-UHFFFAOYSA-N
sodium hydride, sodiumhydride, sodium hydride nah, sodium hydride oil dispersion, sodium monohydride, sodium hydride nad, sodiumhydrid, natrium hydride, sodium hydrid
24758
sodium;hydride
[H-].[Na+]
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
7646-69-7
HNa
23.998
MFCD00003471
BZKBCQXYZZXSCO-UHFFFAOYSA-N
sodium hydride, sodiumhydride, sodium hydride nah, sodium hydride oil dispersion, sodium monohydride, sodium hydride nad, sodiumhydrid, natrium hydride, sodium hydrid
24758
sodium;hydride
[H-].[Na+]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Sodium hydride can be used for direct intercalation into C60, resulting in superconducting material (NaH)4C60. It is primarily used as a strong base in organic synthesis. It can be used to dry some organic solvents. It has been proposed for hydrogen storage for use in fuel cell vehicles.

Solubility
It reacts with water.

Notes
Air and moisture sensitive. Incompatible with oxidizing agents, acid chlorides, acids, halogens and alcohols.
TRUSTED_SUSTAINABILITY
Color Gray to White
Physical Form Moist Powder
Quantity 50 g
UN Number UN1427
Solubility Information It reacts with water.
Formula Weight 24
Odor Odorless
Sensitivity Air and moisture sensitive; Water reactive
Chemical Name or Material Sodium hydride, ≈60% dispersion in oil, in solvent solublebags
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'