missing translation for 'onlineSavingsMsg'
Learn More

Sodium Citrate USP GenAR Crystal, Macron Fine Chemicals™

Catalog Number 15528444
151.05 EUR valid until 2024-12-31
Use promo code "23268" to get your promotional price.


Click to view available options
Glass; 500g
This item is not returnable. View return policy
This item is not returnable. View return policy
CAS 6132-04-3
Packaging Glass
Molecular Formula C6H9Na3O9
Quantity 500 g
Synonym trisodium citrate dihydrate, sodium citrate dihydrate, sodium citrate tribasic dihydrate, natriumcitrat-dihydrat, sodium citrate hydrate, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate, citronensaeure-tri-na-salz-dihydrat, citric acid trisodium salt dihydrate, citric acid, trisodium salt dihydrate, trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate
InChI Key NLJMYIDDQXHKNR-UHFFFAOYSA-K
SMILES C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+]
IUPAC Name trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate
Molecular Weight (g/mol) 294.10
PubChem CID 71474
ChEBI CHEBI:32142
Formula Weight 294.1
Grade USP
Physical Form Crystalline
Chemical Name or Material Sodium Citrate, Dihydrate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'