missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Smoothened Agonist Dihydrochloride Dihydrate

Catalog Number 15426549
Click to view available options
Quantity:
1 mg
5 mg
This item is not returnable. View return policy
364590-63-6
C28H28ClN3OS.2HCl.2H2O
SAG1.3 HCl
This item is not returnable. View return policy
364590-63-6
C28H28ClN3OS.2HCl.2H2O
SAG1.3 HCl

A highly potent activator of Smoothened

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 1 mg
Description Pale yellow
Content And Storage 2°C to 8°C
For Use With (Application) A highly potent activator of Smoothened
Formula Weight 599.00
Sensitivity Light Sensitive and Hygroscopic
Product Type Dihydrochloride Dihydrate
Form Solid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'