Learn More
IKA™ Slot Ring Bearing
An accessory item for the IKA™ S 25N - 8 G Dispersing Tool.
Brand:  IKA™ 1024700
Additional Details : Weight : 0.01000kg
Product Code. 10153383
  • 10.85 € / Each
Estimated Shipment: 13-05-2024
to see stock.
Add to basket
Description
Description
  • Aids in homogenization
  • Supplied individually
Specifications
Specifications
Slot Ring Bearing
IKA™ S 25 N - 8 G Dispersing tool
1 Ea.