missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ SLC39A5 Recombinant Protein
335.00 € - 508.00 €
Specifications
Accession Number | AAH27884 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer |
Gene ID (Entrez) | 283375 |
Molecular Weight (g/mol) | 82.83 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16139853
|
Abnova™
H00283375-P01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16149853
|
Abnova™
H00283375-P01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
AAH27884 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
82.83 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SLC39A5 | |
Human | |
Recombinant | |
Solution |
Antibody Production, Protein Array, ELISA, Western Blot | |
283375 | |
SLC39A5 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VGGSVPNLGPAEQEQNHYLAQLFGLYGENGTLTAGGLARLLHSLGLGRVQGLRLGQHGPLTGRAASPAADNSTHRPQNPELSVDVWAGMPLGPSGWGDLEESKAPHLPRGPAPSGLDLLHRLLLLDHSLADHLNEDCLNGSQLLVNFGLSPAAPLTPRQFALLCPALLYQIDSRVCIGAPAPAPPGDLLSALLQSALAVLLLSLPSPLSLLLLRLLGPRLLRPLLGFLGALAVGTLCGDALLHLLPHAQEGRHAGPGGLPEKDLGPGLSVLGGLFLLFVLENMLGLLRHRGLRPRCCRRKRRNLETRNLDPENGSGMALQPLQAAPEPGAQGQREKNSQHPPALAPPGHQGHSHGHQGGTDITWMVLLGDGLHNLTDGLAIGAAFSDGFSSGLSTTLAVFCHELPHELGDFAMLLQSGLSFRRLLLLSLVSGALGLGGAVLGVGLSLGPVPLTPWVFGVTAGVFLYVALVDMLPALLRPPEPLPTPHVLLQGLGLLLGGGLMLAITLLEERLLPVTTEG | |
LZT-Hs7/MGC34778/ZIP5 | |
SLC39A5 | |
Wheat Germ (in vitro) | |
GST |