missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SIRT6 Synthetic Peptide

Synthetic Peptide

Brand:  Invitrogen™ PEP1081

Additional Details : Weight : 0.01000kg

Product Code. 15295079

  • 130.00 € / 50µg
Estimated Shipment: 12-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


For Research Use Only. All usage must comply with product instructions.


Description

Description

SIRT6 Synthetic Peptide, PEP-1081, from Invitrogen.

This peptide corresponds to 17 amino acids near the carboxy terminus of human SIRT6.PEP-1081 can be used as a blocking peptide with polyclonal antibody PA5-20967.
Specifications

Specifications

51548
purified
Human SIRT6
-20°C
Liquid
Blocking Assay, Control
No
Peptide
RUO - research use only
Human
Q8N6T7
200 μg/mL
Synthetic Peptide
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
SIRT6
0.02% sodium azide
50 μg
PBS
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions