missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG1-Fc Recombinant Human Protein, Invitrogen™ Sino Biological™
This recombinant human protein is expressed from a DNA sequence encoding the human IgG1-Fc region (AAC82527.1), containing residues 99 to 330, including a 103 Cys/Ser mutation
110.00 € - 327.00 €
Specifications
For Use With (Application) | Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories |
---|---|
Molecular Weight (g/mol) | 26kDa |
Storage Requirements | Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. |
Structural Form | Truncated/Recombinant |
Endotoxin Concentration | <1 EU/μg |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
13425946
|
Invitrogen
10702HNAH25 |
5 x 5μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
13455946
|
Invitrogen
10702HNAH50 |
50μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
13435946
|
Invitrogen
10702HNAH250 |
5 x 50μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Immunoglobulin gamma-1 heavy chain constant region, partial (IgG1-Fc), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications.Background: Immunoglobulin G (IgG) is synthesized and secreted by plasma B cells, and constitutes 75% of serum immunoglobulins in humans. IgG antibodies protect the body against the pathogens by agglutination and immobilization, complement activation, toxin neutralization, as well as the antibody-dependent cell-mediated cytotoxicity (ADCC). Recombinant IgG Fc is suggested to represent a potential anti-inflammatory drug for treatment of human autoimmune diseases.
- N-terminal Sequence Analysis: Glu 99
- Activity: 1. Measured by its ability to bind human CD16a-His (Mfr. No. 10389-H08H1) in a functional ELISA; 2. Measured by its ability to bind human CD16a-AVI-His (Mfr. No. 10389-H27H) in a functional ELISA; 3.Measured by its ability to bind human CD16a-His (Mfr. No. 10389-H08H) in a functional ELISA
- Formulation: lyophilized in PBS, pH 7.4, 5% mannitol, 5% trehalose, and 0.02% Tween™-80
- Reconstitution: Dissolve the protein in sterile double-distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
Shipping Condition: Room Temperature. Manufactured by Sino Biological (China).
Specifications
Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories | |
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. | |
<1 EU/μg | |
Fc region of human immunoglobulin G1 | |
IgG1 Fc Tag | |
Growth Factors & Receptors | |
Other Proteins |
26kDa | |
Truncated/Recombinant | |
Fc region of human immunoglobulin G1 | |
IgG1-Fc | |
Mammalian | |
Lyophilized | |
≥95% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.