missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver Nitrate solution 0.1mol/L, Fisher Chemical™
Click to view available options
Quantity:
100 mL
Packaging:
Glass Bottle
Specifications
Concentration | 0.1M (0.1N) |
CAS | 7761-88-8 |
Packaging | Glass Bottle |
Quantity | 100 mL |
Molecular Formula | AgNO3 |
MDL Number | MFCD00003414 |
InChI Key | SQGYOTSLMSWVJD-UHFFFAOYSA-N |
SMILES | [Ag+].[O-][N+]([O-])=O |
IUPAC Name | silver;nitrate |
Molecular Weight (g/mol) | 169.87 |
Show More |
Safety and Handling
Product Identifier
- Silver nitrate 1M Volumetric Solution (SOL 121)
- Danger
- Skin corrosion/irritation Category 1 B
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 2
- H314-Causes severe skin burns and eye damage.
- H410-Very toxic to aquatic life with long lasting effects.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P501-Dispose of contents/container to .
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'