missing translation for 'onlineSavingsMsg'
Learn More

Silver(I) citrate hydrate, Thermo Scientific Chemicals

Catalog Number p-4407841
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
206986-90-5
C6H5Ag3O7
512.70
MFCD00150589
QUTYHQJYVDNJJA-UHFFFAOYSA-K
silver i citrate hydrate, trisilver 1+ citrate hydrate
16212313
2-hydroxypropane-1,2,3-tricarboxylic acid;silver;hydrate
[Ag+].[Ag+].[Ag+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
This item is not returnable. View return policy
206986-90-5
C6H5Ag3O7
512.70
MFCD00150589
QUTYHQJYVDNJJA-UHFFFAOYSA-K
silver i citrate hydrate, trisilver 1+ citrate hydrate
16212313
2-hydroxypropane-1,2,3-tricarboxylic acid;silver;hydrate
[Ag+].[Ag+].[Ag+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

Silver(I) citrate hydrate can be very attractive for the future biomedical or pharmaceutical/therapeutic applications. It can be used as an antimicrobial agent and enhancers of antioxidants and anticancer agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 170°C (decomposition)
Quantity 5 g
Sensitivity Light sensitive
Solubility Information Soluble in water.
Formula Weight 512.70 (Anhydrous)
Physical Form Powder
Chemical Name or Material Silver(I) citrate hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'