missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver Acetate, ReagentPlus™, 99%, Honeywell™
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
Description
- ≥95% for general laboratory use.
Specifications
Color | Beige |
Quantity | 5 g |
Linear Formula | CH3COOAg |
UN Number | UN3077 |
Beilstein | 3595636 |
Formula Weight | 166.91g/mol |
Percent Purity | 99% |
Grade | ReagentPlus™ |
Packaging | Glass Bottle |
Appearance | Powder |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'