Learn More
SHOWA™ 6780R Chemical Resistant Neoprene Gloves, Cotton Liner
Catalog Number 10026111
Click to view available options
12 in. (30 cm) Long; Rough
This item is not returnable. View return policy
This item is not returnable. View return policy
Chemical resistant neoprene glove with soft cotton liner features a rough coating to protect against acids, chemicals and caustics.
  • For use in hot and cold environments
  • Chemical resistant against acids, chemicals and caustics
  • Rough grip ensures precision in wet or greasy environments
  • Soft cotton liner to maintain hand temperature and absorb perspiration
  • Rugged neoprene coating
  • Ideal for applications involving chemicals, engineering, metallurgy, oil/gas and utilities
Certifications/Compliance 21 CFR 170-199
Certifications/Compliance (Europe) Cat III CE 2777, EN 407:X2XXXX, EN 420:2003+A1:2009
Color Black
Latex Free Yes
Material Neoprene
Size 10
EN388 Standard EN 388:2111XP
EN 421 No
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES AKLMPST
EN ISO 374 - 5 Bacteria, Fungi WITH Virus Protection
Glove Cuff Style Gauntlet
Grip Design Rough
Liner Material Cotton
Lining Lined
Model 6780R-10
Series 6780R
Length (English) 12 in.
Length (Metric) 30.5 cm