missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SGK (SGK1), GST Tag Recombinant Protein

Human SGK (SGK1), GST Tag Recombinant Protein

Brand:  Invitrogen™ PR7358A

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15418573

  • 1826.00 € / 100µg
Estimated Shipment: 17-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


For Research Use Only. All usage must comply with product instructions.


Description

Description

SGK (SGK1) may be a transcriptional target for p38/MAPK in hepatoma cells.

Specifications

Specifications

NP_005618, S589D
69.7 kDa
SGK (SGK1)
Human
GST Tag
Ser⁄Thr Kinases: AGC Group
Baculovirus
Recombinant, Catalytic
Serine/Threonine Kinases
Kinase Assay
100μg
SGK Kinase Family
Recombinant
Storage Buffer: 50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Dry Ice
Kinases & Inhibitors
Liquid
Signal Transduction
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions