missing translation for 'onlineSavingsMsg'
Learn More

SERPINB10 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Thermo Scientific PA563751

Additional Details : Weight : 0.02000kg

Product Code. 15751522

  • 514.00 € / 100µL
Estimated Shipment: 11-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Immunogen sequence: INSWVERQTE GKIQNLLPDD SVDSTTRMIL VNALYFKGIW EHQFLVQNTT EKPFRINETT SKPVQMMFMK K Highest antigen sequence identity to the following orthologs: Mouse - 58%, Rat - 79%.

SERPINB10 Polyclonal antibody specifically detects SERPINB10 in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

SERPINB10
Polyclonal
Unconjugated
SERPINB10
9830131G07; BB233602; Bomapin; peptidase inhibitor 10; PI10; PI-10; protease inhibitor 10 (ovalbumin type, bomapin); serine (or cysteine) peptidase inhibitor, clade B (ovalbumin), member 10; serine (or cysteine) peptidase inhibitor, clade B (ovalbumin), member 10, pseudogene; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 10; serpin B10; serpin family B member 10; serpin peptidase inhibitor, clade B (ovalbumin), member 10; SERPINB10; Serpinb10-ps; TGF-beta-repressible serine proteinase inhibitor; transforming growth factor beta repressible serpin; Trespin
Rabbit
Antigen affinity chromatography
RUO
5273
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P48595
SERPINB10
Recombinant protein corresponding to Human SERPINB10
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Research Use Only