missing translation for 'onlineSavingsMsg'
Learn More

SemperIt™ Semperguard™ Latex Innercoated (IC) Powder-Free Gloves

Catalog Number p-8002726
Click to view available options
Size:
X-Small
Medium
This item is not returnable. View return policy
This item is not returnable. View return policy

Manufactured from high quality soft latex film. Semperit™ Semperguard™ Latex Innercoated (IC) Powder-Free Gloves are available in lightly powdered and powder-free styles.

  • Manufactured from high quality soft latex film
  • Powder-free
  • Microroughened outer surface offeres better grip
  • Powder-free gloves have a synthetic inner coating for ease of donning
  • Ambidextrous
  • Rolled cuff
Certifications/Compliance CE Class I, EN 420, EN 374, EN 455, PPE Category III
Coating Material Smooth inside, textured outside
Color Natural
Disposable Single-use
Hand Ambidextrous
Material Rubber
Powdered Powder Free
Size Medium
Sterility Non-sterile
Thickness (Metric) 0.24 mm
For Use With (Application) Suitable for food contact
Glove Cuff Style Rolled
Tensile Strength 18/14 MPa
Length (Metric) 240 mm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'