Learn More
SASS6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA555791
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: QKEQKELQDV GQSLRIKEQE VCKLQEQLEA TVKKLEESKQ LLKNNEKLIT WLNKELNENQ LVRKQDVLGP STTPPAHSSS NTIRSGISPN L Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 82%.
SASS6 Polyclonal antibody specifically detects SASS6 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
SASS6 | |
Polyclonal | |
Unconjugated | |
SASS6 | |
2810453L12Rik; HsSAS-6; MCPH14; RGD1306255; SAS6; SAS-6; SAS-6 centriolar assembly protein; Sass6; spindle assembly 6 homolog; spindle assembly 6 homolog (C. elegans); spindle assembly abnormal protein 6 homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
163786 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6UVJ0 | |
SASS6 | |
Recombinant protein corresponding to Human SASS6 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only