Learn More
ACROBiosystems SARS-CoV-2 (COVID-19) NSP1 Protein, His Tag
Brand:  ACROBiosystems NS1-C51H7-100ug
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16655302
  • 403.51 € / 100µg
Estimated Shipment: 03-01-2022
Add to basket
Description
Description
The viral nonstructural protein 1 (nsP1) is the only membrane-associated protein that anchors the replication complex to the cellular membranes. NSP1 inhibits host translation by interacting with the 40S ribosomal subunit. The nsp1-40S ribosome complex further induces an endonucleolytic cleavage near the 5'UTR of host mRNAs, targeting them for degradation. Viral mRNAs are not susceptible to nsp1-mediated endonucleolytic RNA cleavage thanks to the presence of a 5'-end leader sequence and are therefore protected from degradation. By suppressing host gene expression, nsp1 facilitates efficient viral gene expression in infected cells and evasion from host immune response.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 21.7 kDa. The protein migrates as 22-25 kDa under reducing (R) condition (SDS-PAGE).
Specifications
Specifications
YP_009725297.1
E.coli
SARS-CoV-2
21.7 kDa
100 µg
Met 1 - Gly 180
NSP1,nsp1,Leader protein,Host translation inhibitor nsp1,COVID-19
Unconjugated
PBS, Arginine, pH7.4
Less than 1.0 EU / µg by the LAL method.
Viral Proteins
Powder
>90% as determined by SDS-PAGE
SARS-CoV-2 NSP1, His Tag (NS1-C51H7) is expressed from E.coli cells. It contains AA Met 1 - Gly 180 (Accession # YP_009725297.1).
-20°C
NSP1
His Tag