Learn More
ACROBiosystems SARS-CoV-2 (COVID-19) NSP1 Protein, His Tag
483.87 € - 3495.90 €
Specifications
Accession Number | YP_009725297.1 |
---|---|
Molecular Weight (g/mol) | 21.7 kDa |
Source | SARS-CoV-2 NSP1, His Tag (NS1-C51H7) is expressed from E.coli cells. It contains AA Met 1 - Gly 180 (Accession # YP_009725297.1). |
Immunogen | Met 1 - Gly 180 |
Storage Requirements | -20°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16665302
|
ACROBiosystems
NS1-C51H7-1MG |
1 mg |
3495.90 €
1mg |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16655302
|
ACROBiosystems
NS1-C51H7-100UG |
100 μg |
483.87 €
100µg |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The viral nonstructural protein 1 (nsP1) is the only membrane-associated protein that anchors the replication complex to the cellular membranes. NSP1 inhibits host translation by interacting with the 40S ribosomal subunit. The nsp1-40S ribosome complex further induces an endonucleolytic cleavage near the 5'UTR of host mRNAs, targeting them for degradation. Viral mRNAs are not susceptible to nsp1-mediated endonucleolytic RNA cleavage thanks to the presence of a 5'-end leader sequence and are therefore protected from degradation. By suppressing host gene expression, nsp1 facilitates efficient viral gene expression in infected cells and evasion from host immune response.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 21.7 kDa. The protein migrates as 22-25 kDa under reducing (R) condition (SDS-PAGE).Specifications
YP_009725297.1 | |
SARS-CoV-2 NSP1, His Tag (NS1-C51H7) is expressed from E.coli cells. It contains AA Met 1 - Gly 180 (Accession # YP_009725297.1). | |
-20°C | |
NSP1 | |
SARS-CoV-2 | |
PBS, Arginine, pH7.4 | |
E.coli | |
Viral Proteins |
21.7 kDa | |
Met 1 - Gly 180 | |
NSP1,nsp1,Leader protein,Host translation inhibitor nsp1,COVID-19 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |