Learn More
Greiner Bio-One Sapphire Polypropylene Full-Skirted 384-well PCR Microplates
Suitable for adhesive sealers and heat sealing
Brand: Greiner Bio-One 785201
Additional Details : Weight : 0.80000kg
Description
Blue alphanumeric well codingSpecifications
Standard | |
Not Autoclavable | |
384 |
16 x 24 | |
Polypropylene | |
PCR |