Learn More
Roetzmeier™ Safety Transportation Canister with 1.5 in. Screw Cap
High-quality safety canister designed to safely transport solvents.
Brand: Roetzmeier™ 10KT
Additional Details : Weight : 0.10000kg
Description
- Made of stainless steel
- Gaskets made of PTFE
- Flame trap in opening
- Conductive – can be bonded using any part of the canister
Specifications
UN | |
Screw Cap | |
10 L | |
Stainless Steel | |
Flame Trap and Screw Cap | |
Rectangular | |
0.8 mm | |
15 in. | |
1.6 in. | |
5.5 lb. |
Yes | |
Stainless Steel w/PTFE Gasket | |
D150, A15, TR2, TR2V | |
Electro-polished | |
200°C | |
0.032 in. | |
130 x 280 x 380 mm | |
380 mm | |
40 mm | |
2.501 Kg |