Learn More
(S)-(-)-N-(3-Pentyl)-1-phenylethylamine hydrochloride, 99%, Thermo Scientific™
Product Code. 11334975
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 11334975
Brand: Thermo Scientific Alfa Aesar L18357.03
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 374790-92-8
Molecular Formula C13H22ClN
Molecular Weight (g/mol) 227.78
MDL Number MFCD06199157
InChI Key RKPRPVRHVBFULB-UHFFFAOYNA-N
Synonym s---n-3-pentyl-1-phenylethylamine hydrochloride, s-n-1-phenylethyl pentan-3-amine hydrochloride, pentan-3-yl 1s-1-phenylethyl amine hydrochloride
PubChem CID 22831494
IUPAC Name N-[(1S)-1-phenylethyl]pentan-3-amine;hydrochloride
SMILES Cl.CCC(CC)NC(C)C1=CC=CC=C1
Specifications
Melting Point 232°C to 234°C
Quantity 1 g
Sensitivity Hygroscopic
Formula Weight 227.78
Percent Purity 99%
Chemical Name or Material (S)-(-)-N-(3-Pentyl)-1-phenylethylamine hydrochloride