missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-1-Methyl-3-hydroxypyrrolidine, 98%, ee 99%, Thermo Scientific™

Catalog Number p-7047739
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
104641-59-0
C5H11NO
101.15
MFCD03788748
FLVFPAIGVBQGET-UHFFFAOYNA-N
s-+-1-methyl-3-pyrrolidinol, 3s-1-methylpyrrolidin-3-ol, s-1-methyl-3-pyrrolidinol, s-1-methylpyrrolidin-3-ol, 3-pyrrolidinol, 1-methyl-, 3s, s-+-3-hydroxy-n-methylpyrrolidine, s-+-1-methyl-3-hydroxypyrrolidine, s-3-hydroxy-1-methylpyrrolidine, 1-methyl-3-pyrrolidinol, +, s-3-hydroxy-1-methyl-pyrrolidine
643504
(3S)-1-methylpyrrolidin-3-ol
CN1CCC(O)C1
This item is not returnable. View return policy
104641-59-0
C5H11NO
101.15
MFCD03788748
FLVFPAIGVBQGET-UHFFFAOYNA-N
s-+-1-methyl-3-pyrrolidinol, 3s-1-methylpyrrolidin-3-ol, s-1-methyl-3-pyrrolidinol, s-1-methylpyrrolidin-3-ol, 3-pyrrolidinol, 1-methyl-, 3s, s-+-3-hydroxy-n-methylpyrrolidine, s-+-1-methyl-3-hydroxypyrrolidine, s-3-hydroxy-1-methylpyrrolidine, 1-methyl-3-pyrrolidinol, +, s-3-hydroxy-1-methyl-pyrrolidine
643504
(3S)-1-methylpyrrolidin-3-ol
CN1CCC(O)C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Density 0.92
Boiling Point 103°C to 106°C (35 mmHg)
Flash Point 70°C (158°F)
Refractive Index 1.464
Quantity 250 mg
Beilstein 4349394
Sensitivity Air sensitive; Hygroscopic
Formula Weight 101.15
Percent Purity 98%
Assay ee 99%
Chemical Name or Material (S)-(+)-1-Methyl-3-hydroxypyrrolidine
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'