missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPS6KB2 (p70S6K Beta) Recombinant Human Protein
RPS6KB2 (p70S6K Beta) Protein
Brand: Invitrogen PV3831
Additional Details : Weight : 0.05250kg
Description
RPS6KB2 (p70S6K beta) is a serine⁄threonine kinase with 2 non-identical kinase catalytic domains which phosphorylates the S6 ribosomal protein and eukaryotic translation initiation factor 4B (eIF4B). Phosphorylation of S6 leads to an increase in protein synthesis and cell proliferation.Specifications
NP_003943 | |
Cell Viability, Pharma and Biopharma, Cell Proliferation and Function, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
82.2kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
RPS6KB2 (p70S6K Beta) | |
RPS6KB2 (p70S6K beta) was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β−glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 100μM ATP, 200μg/mL S6 peptide substrate (AKRRRLSSLRA) and trace [32P]-γ-ATP for 10 minutes at 30°C | |
RPS6KB2 (p70S6K Beta) Recombinant Human Protein | |
Human | |
Baculovirus | |
Serine/threonine kinase |
0.24mg/mL | |
114294, 6199 | |
RPS6KB2 (p70S6K Beta) Recombinant Human Protein | |
–80°C | |
1 Each | |
RPS6KB2 (p70S6K Beta) Recombinant Human Protein | |
RSK (RPS6K) Kinase family | |
40nmoles of phosphate transferred to S6 peptide substrate (AKRRRLSSLRA) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 16.7μg/mL | |
GST Tag | |
Kinases and inhibitors | |
95% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.