missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human RILPL2 Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP1402
Additional Details : Weight : 0.01000kg
Description
RLP2 Synthetic Peptide, PEP-1402, from Invitrogen.
This peptide corresponds to 16 amino acids near the center of human RLP2.PEP-1402 can be used as a blocking peptide with polyclonal antibody PA5-34359.Specifications
196383 | |
purified | |
Human RILPL2 | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
Q969X0 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
RILPL2 | |
0.02% sodium azide | |
50 μg | |
PBS |