Learn More
Riociguat, MedChemExpress
Product Code. 16476546
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
Product Code. 16476546
Brand: Medchem Express HY14779
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Riociguat is an oral stimulator of soluble guanylate cyclase (sGC) used in the treatment of pulmonary hypertension.
Riociguat is an oral stimulator of soluble guanylate cyclase (sGC) used in the treatment of pulmonary hypertension.
Specifications
Chemical Name or Material Riociguat
CAS 625115-55-1
Color Yellow
Purity Grade Notes Research
Molecular Formula C20H19FN8O2
Quantity 100 mg
Synonym BAY 632521
Solubility Information DMSO : 20 mg/mL (47.35 mM; Need ultrasonic)
SMILES FC1=CC=CC=C1CN2C3=NC=CC=C3C(C4=NC(N)=C(C(N)=N4)N(C)C(OC)=O)=N2
Molecular Weight (g/mol) 422.42
Formula Weight 422.42
Percent Purity 95.1%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation