missing translation for 'onlineSavingsMsg'
Learn More

Rhodium, plasma standard solution, Specpure™ Rh 1000μg/mL, Thermo Scientific Chemicals

Catalog Number p-4782100
Click to view available options
Quantity:
50 mL
100 mL
500 mL
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7440-16-6
Rh
102.91
MFCD00011201 MFCD03458392
MHOVAHRLVXNVSD-UHFFFAOYSA-N
23948
CHEBI:33359
rhodium
[Rh]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7440-16-6
Rh
102.91
MFCD00011201 MFCD03458392
MHOVAHRLVXNVSD-UHFFFAOYSA-N
23948
CHEBI:33359
rhodium
[Rh]

Rhodium, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 50 mL
Odor Characteristic
Solubility Miscible with water
Physical Form Liquid
Chemical Name or Material Rhodium, Rh 1000μg/mL

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'