Learn More
Thermo Scientific™ Remel™ Brucella Broth (Dehydrated)
Cultivate Brucella spp. and other fastidious microorganisms using Brucella Broth (Dehydrated).
Brand:  Thermo Scientific™ R452662
Product Code. 11942902
Description
Description
Cultivate Brucella species and a variety of microorganisms using Thermo Scientific™ Remel™ Brucella Broth (Dehydrated). Brucella Broth prepared according to the formula of Albimi Broth is recommended by the American Public Health Association1. Albimi Broth is used to isolate Brucella from dairy products, and foods2. Brucella Broth is used for preliminary enrichment of Brucella and it is isolated using Brucella Broth in a biphasic medium.
This medium contains casein and meat peptones which provide nitrogen, amino acids and peptides necessary for bacterial growth. Sodium bisulfite is a reducing agent. Yeast extract is a source of B-complex vitamins. Sodium chloride is a source of essential electrolytes and maintains osmotic equilibrium. Dextrose provides a ready source of energy.
  • Easy to prepare - Convenient to use
  • Easy to store - No special storage required
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Industrial Reference: USDA.
Powder
500 g
Brucella Broth
Dehydrated Culture Media