Learn More
Trajan Scientific and Medical™ Reducing Unions
Gives low dead volume connections for optimum chromatography.
59.60 €
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
15219103
|
Trajan Scientific and Medical™
101290 |
Size: 1/8 in. to 1/16 in. |
59.60 €
Each |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
15299093
|
Trajan Scientific and Medical™
101230 |
Size: 1/4 in. to 1/16 in. |
59.60 €
Each |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
Description
- Low dead volume
- High temperature applications
- Leak free connections
Specifications
Reducing Union |