Learn More
Trajan Scientific and Medical™ Reducing Unions
Gives low dead volume connections for optimum chromatography.
Brand: Trajan Scientific and Medical™ 101290
Additional Details : Weight : 0.01000kg
Description
- Low dead volume
- High temperature applications
- Leak free connections
Specifications
Reducing Union | |
Size: 1/8 in. to 1/16 in. |