Learn More
Trajan Scientific and Medical™ Reducing Unions
Gives low dead volume connections for optimum chromatography.
Brand:  Trajan Scientific and Medical™ 101290
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15219103
  • 59.60 € / Each
Estimated Shipment: 13-05-2024
to see stock.
Add to basket
Description
Description
  • Low dead volume
  • High temperature applications
  • Leak free connections
Specifications
Specifications
Reducing Union
Size: 1/8 in. to 1/16 in.