missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Sheep TNF-alpha Protein
A DNA sequence encoding the Ovis aries (Sheep) TNF-alpha, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6812-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001020031.1 | |
TNF-alpha Protein | |
Research Use Only | |
TNF | |
Recombinant Protein | |
E. coli | |
LRSSSQASNN KPVAHVVANI SAPGQLRWGD SYANALMANG VELKDNQLVV PTDGLYLIYS QVLFRGHGCP STPLFLTHTI SRIAVSYQTK VNILSAIKSP CHRETLEGAE AKPWYEPIYQ GGVFQLEKGD RLSAEINLPE YLDYAESGQV YFGIIAL | |
Greater than 90% as determined by SDS-PAGE. |
443540 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Sheep | |
GST | |
Tris-based buffer, 50% glycerol |