missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Sheep Interleukin-8 Protein

A DNA sequence encoding the Ovis aries (Sheep) Interleukin-8, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6228-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16071760

  • 920.10 € / 100µg

Estimated Shipment: 10-01-2022
Add to basket

Description

Description

Specifications

Specifications

NP_001009401.1
Interleukin-8 Protein
100 μg
Not determined.
Full Length
Sheep
His
Tris-based buffer, 50% glycerol
443418
Greater than 90% as determined by SDS-PAGE.
Research Use Only
IL8
Recombinant Protein
E. coli
AVLSRMSTEL RCQCIKTHST PFHPKFIKEL RVIESGPHCE NSEIIVKLTN GKEVCLDPKE KWVQKVVQAF LKRAEKQDP
Videos
SDS
Documents
Certificates
Promotions

Promotions